missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1orf107 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21076-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
INTS5 is a subunit of the Integrator complex, which associates with the C-terminal domain of RNA polymerase II large subunit.Specifications
C1orf107 | |
Polyclonal | |
Unconjugated | |
DIEXF | |
C1orf107, DEF | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27042 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q68CQ4 | |
DIEXF | |
C1orf107 Fusion Protein Ag15250 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |