missing translation for 'onlineSavingsMsg'
Learn More

C1orf113 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 25767-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16824335

  • 1236.20 SEK / 20µL
Estimated Shipment: 08-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

C1orf113
0.5 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
A4FU49, Q7TSG5
C1orf113, cDNA FLJ58582, Homo sapiens (Human), SH3D21
Rabbit
IgG
20 μL
-20°C
Primary
66938, 79729
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
SH3D21
Liquid
Sh3d21
C1orf113 Fusion Protein Ag22643
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Videos
SDS
Documents
Certificates
Special Offers

Special Offers