missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1orf92 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | C1orf92 |
---|---|
Concentration | 0.24 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16850625
|
Proteintech
21031-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 30-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16840625
|
Proteintech
21031-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 30-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
LRRC71 is a protein coding gene.Specifications
C1orf92 | |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q8N4P6 | |
149499 | |
C1orf92 Fusion Protein Ag15344 | |
Primary | |
-20°C | |
LRRC71 |
0.24 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
C1orf92, LRRC71, RP11 356J7.1 | |
LRRC71 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |