missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1QTNF3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11414-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Adipose tissue of an organism plays a major role in regulating physiologic and pathologic processes such as metabolism and immunity by producing and secreting a variety of bioactive molecules termed adipokines. One highly conserved family of adipokines is adiponectin/ACRP30 and its structural and functional paralogs, the C1q/tumor necrosis factor-alpha-related proteins (CTRPs) 1-7. Unlike adiponectin, which is expressed exclusively by differentiated adipocytes, the CTRPs are expressed in a wide variety of tissues. An analysis of the crystal structure of adiponectin revealed a structural and evolutionary link between TNF and C1q-containing proteins, suggesting that these proteins arose from a common ancestral innate immunity gene. Multiple isoforms of human CTRP3 have been reported. It has been suggested that CTRP3 may play a role in skeletal development.Specifications
C1QTNF3 | |
Polyclonal | |
Unconjugated | |
C1QTNF3 | |
C1ATNF3, C1QTNF3, Corcs, Cors, CORS26, CTRP3, Secretory protein CORS26 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
114899, 294806, 81799 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9BXJ4, Q9ES30 | |
C1qtnf3 | |
C1QTNF3 Fusion Protein Ag1958 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |