Läs mer
C20 Ceramide, TRC
Produktkod. 30362492
Change view
Kvantitet:
5 mg
25 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30362492 25 mg
30364353 250 mg
30353881 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30362492 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC26312025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 7344-02-7
Molekylformel C38 H75 N O3
Molekylvikt (g/mol) 594.01
IUPAC-namn N-[(E,2R,3S)-1,3-dihydroxyoctadec-4-en-2-yl]icosanamide
LEDER CCCCCCCCCCCCCCCCCCCC(=O)N[C@H](CO)[C@@H](O)\C=C\CCCCCCCCCCCCC
Specifikationer
Kemiskt namn eller material C20 Ceramide
Kvantitet 25 mg
InChI formel InChI=1S/C38H75NO3/c1-3-5-7-9-11-13-15-17-18-19-20-22-24-26-28-30-32-34-38(42)39-36(35-40)37(41)33-31-29-27-25-23-21-16-14-12-10-8-6-4-2/h31,33,36-37,40-41H,3-30,32,34-35H2,1-2H3,(H,39,42)/b33-31+/t36-,37+/m1/s1
Formel vikt 593.5747