missing translation for 'onlineSavingsMsg'
Learn More

C20orf19 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 21177-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16850795

  • 1656.73 SEK / 20µL
Estimated Shipment: 10-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Centrosomal protein required for establishing a robust mitotic centrosome architecture that can endure the forces that converge on the centrosomes during spindle formation. Required for stabilizing the expanded pericentriolar material around the centriole.
Specifications

Specifications

C20orf19
Polyclonal
Unconjugated
KIZ
C20orf19, Centrosomal protein kizuna, HT013, KIZ, NCRNA00153, non protein coding RNA 153, PLK1S1, Polo like kinase 1 substrate 1
Rabbit
Antigen Affinity Chromatography
RUO
55857
-20°C
Liquid
Immunocytochemistry, Western Blot, Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin)
0.51 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q2M2Z5
KIZ
C20orf19 Fusion Protein Ag15454
20 μL
Primary
Human
Antibody
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales