missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C20orf7 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26359-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The NADH-ubiquinone oxidoreductase complex (complex I) of the mitochondrial respiratory chain catalyzes the transfer of electrons from NADH to ubiquinone, and consists of at least 43 subunits. The complex is located in the inner mitochondrial membrane. This gene encodes a mitochondrial protein that is associated with the matrix face of the mitochondrial inner membrane and is required for complex I assembly. A mutation in this gene results in mitochondrial complex I deficiency. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
C20orf7 | |
Polyclonal | |
Unconjugated | |
NDUFAF5 | |
C20orf7, NDUFAF5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
79133 | |
-20°C | |
Liquid |
Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5TEU4 | |
NDUFAF5 | |
C20orf7 Fusion Protein Ag23822 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |