missing translation for 'onlineSavingsMsg'
Learn More

C2orf79/PTRHD1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 25576-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16824135

  • 1236.20 SEK / 20µL
Estimated Shipment: 30-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

C2orf79/PTRHD1
0.4 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q6GMV3
C2orf79, PTRHD1
Rabbit
IgG
20 μL
-20°C
Primary
391356
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
PTRHD1
Liquid
PTRHD1
C2orf79/PTRHD1 Fusion Protein Ag22255
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
SDS
Documents
Certificates
Special Offers

Special Offers