Läs mer
C3orf67 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-57722
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
C3orf67 Polyclonal specifically detects C3orf67 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifikationer
| C3orf67 | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunocytochemistry/Immunofluorescence 1-4 ug/ml | |
| Chromosome 3 Open Reading Frame 67 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 200844 | |
| Human | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| C3orf67 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:IDLVAFTSEIFKGAVFQSLDGIVVSANCKLRKIFTLKSKPQDTADKDAVYGVPFSTDEPTDIIPRSCQLMTDVPHVTQLLNMTKLRQTEIKFGGHPLRSAE | |
| 100 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Korrigering av produktinnehåll
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering