missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ C4BPA (Human) Recombinant Protein
Human C4BPA full-length ORF ( AAH22312, 49 a.a. - 597 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00000722-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: NCGPPPTLSFAAPMDITLTETRFKTGTTLKYTCLPGYVRSHSTQTLTCNSDGEWVYNTFCIYKRCRHPGELRNGQVEIKTDLSFGSQIEFSCSEGFFLIGSTTSRCEVQDRGVGWSHPLPQCEIVKCKPPPDIRNGRHSGEENFYAYGFSVTYSCDPRFSLLGHASISCTVENETIGVWRPSPPTCEKITCRKPDVSHGEMVSGFGPIYNYKDTIVFKCQKGFVLRGSSVIHCDADSKWNPSPPACEPNSCINLPDIPHASWETYPRPTKEDVYVVGTVLRYRCHPGYKPTTDEPTTVICQKNLRWTPYQGCEALCCPEPKLNNGEITQHRKSRPANHCVYFYGDEISFSCHETSRFSAICQGDGTWSPRTPSCGDICNFPPKIAHGHYKQSSSYSFFKEEIIYECDKGYILVGQAKLSCSYSHWSAPAPQCKALCRKPELVNGRLSVDKDQYVEPENVTIQCDSGYGVVGPQSITCSGNRTWYPEVPKCEWETPEGCEQVLTGKRLMQCLPNPEDVKMALEVYKLSLEIEQLELQRDSARQSTLDKEL
Specifications
AAH22312 | |
complement component 4 binding protein, alpha | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C4BPA | |
GST |
722 | |
Wheat germ expression system | |
10 μg | |
C4BP, PRP | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |