missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ C4BPB (Human) Recombinant Protein
Human C4BPB full-length ORF ( AAH05378, 1 a.a. - 251 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00000725-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MFFWCACCLMVAWRVSASDEHCPELPPVDNSIFVAKEVEGQILGTYVCIKGYHLVGKKTLFCNASKEWDNTTTECRLGHCPDPVLVNGEFSSSGPVNVSDKITFMCNDHYILKGSNRSQCLEDHTWAPPFPICKSRDCDPPGNPVHGYFEGNNFTLGSTISYYCEDRYYLVGVQEQQCVDGEWSSALPVCKLIQEAPKPECEKALLAFQESKNLCEAMENFMQQLKESGMTMEELKYSLELKKAELKAKLL
Specifications
AAH05378 | |
complement component 4 binding protein, beta | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C4BPB | |
GST |
725 | |
Wheat germ expression system | |
25 μg | |
C4BP | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |