missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C4orf29 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25036-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
C4orf29 (Chromosome4 open reading frame 29)is a secretedproteinin humans that is encoded by the C4orf29genewhich maps to human chromosome 4. The function of C4orf29 is uncharacterized.Specifications
C4orf29 | |
Polyclonal | |
Unconjugated | |
ABHD18 | |
C4orf29 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
269423, 80167 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.43 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q0P651, Q8C1A9 | |
Abhd18 | |
C4orf29 Fusion Protein Ag19120 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |