missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C9orf86 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20848-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Maternal embryonic leucine-zipper kinase (MELK) is a key regulator of survival of stemlike GBM cells in vitro. MELK expression is increased in breast cancer tissue and this increase is also associated with poor patient survival, as predicted for a candidate oncogene.Specifications
C9orf86 | |
Polyclonal | |
Unconjugated | |
RABL6 | |
C9orf86, Parf, Partner of ARF, pp8875, RBEL1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55684 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q3YEC7 | |
RABL6 | |
C9orf86 Fusion Protein Ag14350 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |