missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C9orf9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | C9orf9 |
---|---|
Concentration | 0.26 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16824565
|
Proteintech
26034-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16814565
|
Proteintech
26034-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Product No. 26034-1-AP is a rabbit polyclonal antibody raised against the full-length of human C9orf9. Two isoforms were produced by alternative splicing with predicted MW of 19 and 26 kDa. Two bands at 16 kDa and 30 kDa could be detected using the presen
SPACA9 (Sperm Acrosome Associated 9) is a Protein Coding gene.Specifications
C9orf9 | |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q7TPM5, Q96E40 | |
11092, 296610, 69987 | |
C9orf9 Fusion Protein Ag23320 | |
Primary | |
-20°C | |
SPACA9 |
0.26 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
C9orf9, Uncharacterized protein C9orf9 | |
Spaca9 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |