missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C9orf98 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21036-1-AP-20UL
Additional Details : Weight : 0.00010kg
Specifications
C9orf98 | |
Polyclonal | |
Unconjugated | |
AK8 | |
C9orf98, DDX31, RP11 143F18.1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
158067 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96MA6 | |
AK8 | |
C9orf98 Fusion Protein Ag15358 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |