Läs mer
Tocris Bioscience™ CaCCinh-A01
GREENER_CHOICE
Produktkod. 30001580
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30001580 10 mg
30001600 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30001580 Leverantör Tocris Bioscience™ Leverantörsnummer 4877/10
Calcium-activated chloride channel (CaCC) inhibitor
Calcium-activated chloride channel (CaCC) inhibitor (IC50 ~ 10 μM). Inhibits CaCC currents in human bronchial and intestinal cells. Also inhibits TMEM16A channels (IC50 = 2.1 μM, in TMEM16A-expressing FRT cells).
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 407587-33-1
Molekylformel C18H21NO4S
Specifikationer
Kemiskt namn eller material CaCCinh-A01
Kvantitet 10 mg
Formel vikt 347.43
Renhet >99%
Molekylvikt (g/mol) 347.429