Läs mer
Cadaverine-15N2 Dihydrochloride, TRC
Produktkod. 30137759
Change view
Kvantitet:
2.5 mg
25 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30137759 25 mg -
30009547 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30137759 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC05800225MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2747-91-3
Molekylformel C5 H14 15N2
Molekylvikt (g/mol) 104.165
Synonym 1,5-Pentanediamine-15N2 Dihydrochloride, 1,5-Amylene Diamine-15N2 Dihydrochloride, 1,5-Diamino-n-pentane, 1,5-Diaminopentane-15N2 Dihydrochloride, 1,5-Pentamethylenediamine-15N2 Dihydrochloride, Pentamethylenediamine-15N2 Dihydrochloride, α,ω-Pentanediamine-15N2 Dihydrochloride
IUPAC-namn pentane-1,5-di(^{15}N_{2})amine
LEDER [15NH2]CCCCC[15NH2]
Specifikationer
Kemiskt namn eller material Cadaverine-15N2 Dihydrochloride
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C5H14N2/c6-4-2-1-3-5-7/h1-7H2/i6+1,7+1
Formel vikt 104.11
Procent renhet >95%