missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CADM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14335-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Mediates homophilic cell-cell adhesion in a Ca (2+)-independent manner. Also mediates heterophilic cell-cell adhesion with CADM3 and PVRL3 in a Ca (2+)-independent manner. Acts as a tumor suppressor in non-small-cell lung cancer (NSCLC) cells. Interaction with CRTAM promotes natural killer (NK) cell cytotoxicity and interferon-gamma (IFN-gamma) secretion by CD8+ cells in vitro as well as NK cell-mediated rejection of tumors expressing CADM3 in vivo. May contribute to the less invasive phenotypes of lepidic growth tumor cells. In mast cells, may mediate attachment to and promote communication with nerves. CADM1, together with MITF, is essential for development and survival of mast cells in vivo. May act as a synaptic cell adhesion molecule that drives synapse assembly. May be involved in neuronal migration, axon growth, pathfinding, and fasciculation on the axons of differentiating neurons. May play diverse roles in the spermatogenesis including in the adhesion of spermatocytes and spermatids to Sertoli cells and for their normal differentiation into mature spermatozoa.Specifications
CADM1 | |
Polyclonal | |
Unconjugated | |
CADM1 | |
BL2, CADM1, cell adhesion molecule 1, IGSF4, IGSF4A, Necl 2, NECL2, Nectin like protein 2, RA175, sgIGSF, ST17, sTSLC 1, SYNCAM, synCAM1, TSLC 1, TSLC1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23705, 363058, 54725 | |
-20°C | |
Liquid |
Western Blot | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6AYP5, Q8R5M8, Q9BY67 | |
Cadm1 | |
CADM1 Fusion Protein Ag5621 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |