Läs mer
Tocris Bioscience™ Caerulein
GREENER_CHOICE
CCK agonist
Brand:  Tocris Bioscience™ 6264/1
Encompass
Produktkod. 15746835
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter »
Beskrivning
Beskrivning
Caerulein is a CCK agonist. Stimulates biosynthesis of pancreatic enzymes in AR4-2J pancreatic cells in vitro. Induces acute pancreatitis in mice. Also enhances proteasome activity in Gdeg mice.
Specifikationer
Specifikationer
Human
Store at −20°C
1 mg
XQDYTGWMDF
17650-98-5
1352.4g/mol
RUO - research use only
Produktförslag
Produktförslag
Filmer
SDS
Dokument
Dokument
Produktspecifikationer
Kampanjer
Kampanjer