Läs mer
Caffeic Acid Benzyl Ester, TRC
Produktkod. 30339664
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30339664 10 mg
1 options
Produktkod. 30339664 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC08000510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Caffeic Acid Benzyl Ester
CAS 107843-77-6
Molekylformel C16 H14 O4
Kvantitet 10 mg
Synonym 3-(3,4-Dihydroxyphenyl)-2-propenoic Acid Phenylmethyl Ester, 3-(3,4-Dihydroxyphenyl)-2-propenoic Acid Benzyl Ester, Benzyl Caffeate, NSC 666590
InChI formel InChI=1S/C16H14O4/c17-14-8-6-12(10-15(14)18)7-9-16(19)20-11-13-4-2-1-3-5-13/h1-10,17-18H,11H2/b9-7+
LEDER Oc1ccc(\C=C\C(=O)OCc2ccccc2)cc1O
IUPAC-namn benzyl (E)-3-(3,4-dihydroxyphenyl)prop-2-enoate
Molekylvikt (g/mol) 270.28
Formel vikt 270.0892