Läs mer
Caffeic Acid Methyl Ester, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Caffeic Acid Methyl Ester |
| CAS | 67667-67-8 |
| Molekylformel | C10 H10 O4 |
| Kvantitet | 250 mg |
| Synonym | 2-Propenoic acid, 3-(3,4-dihydroxyphenyl)-, methyl ester, (2E)-, 2-Propenoic acid, 3-(3,4-dihydroxyphenyl)-, methyl ester, (E)-, (E)-3-(3,4-Dihydroxyphenyl)propenoic acid methyl ester, (E)-Caffeic acid methyl ester, Methyl (E)-3-(3,4-dihydroxyphenyl)acrylate, trans-Caffeic acid methyl ester, trans-3,4-Dihydroxycinnamic acid methyl ester, Methyl trans-3-(3,4-dihydroxyphenyl)acrylate, Methyl (2E)-3-(3,4-dihydroxyphenyl)-2-propenoate |
| InChI formel | InChI=1S/C10H10O4/c1-14-10(13)5-3-7-2-4-8(11)9(12)6-7/h2-6,11-12H,1H3/b5-3+ |
| LEDER | COC(=O)\C=C\c1ccc(O)c(O)c1 |
| IUPAC-namn | methyl (E)-3-(3,4-dihydroxyphenyl)prop-2-enoate |
| Molekylvikt (g/mol) | 194.18 |
| Formel vikt | 194.0579 |