Läs mer
Caffeine-13C, TRC
Produktkod. 30004089
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30004089 10 mg
30005131 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30004089 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC08010310MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 202282-98-2
Molekylformel C713CH10N4O2
Molekylvikt (g/mol) 195.18
Synonym 3,7-Dihydro-1,7-dimethyl-3-(methyl-13C)-1H-Purine-2,6-dione
IUPAC-namn 1,7-dimethyl-3-(113C)methylpurine-2,6-dione
LEDER O=C(N1C)C(N2C)=C(N=C2)N([13CH3])C1=O
Specifikationer
Kemiskt namn eller material Caffeine-13C
Kvantitet 10 mg
InChI formel InChI=1S/C8H10N4O2/c1-10-4-9-6-5(10)7(13)12(3)8(14)11(6)2/h4H,1-3H3/i2+1