Learn More
Merck Millipore Calbiochem™ Smoothened Agonist, SAG
A cell-permeable Smoothened Agonist, SAG, CAS 364590-63-6, modulates the coupling of Smo with its downstream effector by interacting with the Smo heptahelical domain (KD = 59 nM).
Brand:  Merck Millipore 566660-1MG
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 15340394
  • 2360.00 SEK / 1mg
Estimated Shipment
02-11-2020
Add to basket
Description
Description
Packaged Under Inert Gas
Specifications
Specifications
98% by HPLC
Hygroscopic
Protect from Light
364590-63-6
599g/mol
Light yellow solid
1mg
C28H28ClN3OS·2HCl·2H2O
DMSO or H2O