Learn More
Calcein, MP Biomedicals™
Product Code. 11474622 Shop All MP Biomedicals Products
Change view
Click to view available options
Kvantitet:
1 g
5 g
10 g
25 g
Unit Size:
10g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet unitSize
11474622 25 g 25g
11444622 1 g 1g
11454622 5 g 5g
11464622 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11474622 Supplier MP Biomedicals Supplier No. 0219016725
Chemical Identifiers
CAS 1461-15-0
Molekylformel C30H26N2O13
Molekylvikt (g/mol) 622.539
InChI-nyckel DEGAKNSWVGKMLS-UHFFFAOYSA-N
Synonym Fluorexon, Calcein W
PubChem CID 65079
ChEBI CHEBI:51903
IUPAC-namn 2-[[7'-[[bis(karboximetyl)amino]metyl]-3',6'-dihydroxi-3-oxospiro[2-bensofuran-1,9'-xanten]-2'-yl]metyl-(karboximetyl)amino]ättiksyra
LEDER C1=CC=C2C(=C1)C(=O)OC23C4=CC(=C(C=C4OC5=CC(=C(C=C35)CN(CC(=O)O)CC(=O)O)O)O)CN(CC(=O)O)CC(=O)O
Specifications
Kemiskt namn eller material Calcein
Namnnotering Suitable for fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.
Formel vikt 622.5
Färg Orange
Kvantitet 25 g
Fysisk form Pulver