Läs mer
Calcium 3-methyl-2-oxopentanoate, TRC
Produktkod. 30364461
Change view
Kvantitet:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30364461 100 mg
30363865 50 mg
30362399 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30364461 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC280040100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 66872-75-1
Molekylformel 2 C6 H9 O3 . Ca
Molekylvikt (g/mol) 298.35
Synonym Pentanoic acid, 3-methyl-2-oxo-, calcium salt (2:1), Pentanoic acid, 3-methyl-2-oxo-, calcium salt (9CI), Pentanoic acid, 3-methyl-2-oxo-, calcium salt, (±)-, Calcium 3-methyl-2-oxopentanoate, calcium bis(3-methyl-2-oxopentanoate), α-keto-Isoleucine calcium, 3-Methyl-2-oxopentanoic acid calcium salt, Calcium (±)-3-methyl-2-oxovalerate
IUPAC-namn calcium;3-methyl-2-oxopentanoate
LEDER [Ca+2].CCC(C)C(=O)C(=O)[O-].CCC(C)C(=O)C(=O)[O-]
Specifikationer
Kemiskt namn eller material 3-Methyl-2-oxovaleric acid calcium salt
Kvantitet 100 mg
InChI formel InChI=1S/2C6H10O3.Ca/c2*1-3-4(2)5(7)6(8)9;/h2*4H,3H2,1-2H3,(H,8,9);/q;;+2/p-2
Formel vikt 298.0729