Läs mer
Calcium a-Hydroxy-gamma-Methylmercaptobutyrate, TRC
Produktkod. 30312916
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30312916 500 mg
1 options
Produktkod. 30312916 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA422097500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Hydroxy-4-(methylthio)butanoic Acid Calcium Salt
CAS 4857-44-7
Anteckningar om renhetsgrad HPLC
Molekylformel 2 C5 H9 O3 S . Ca
Kvantitet 500 mg
Synonym Butanoic acid, 2-hydroxy-4-(methylthio)-, calcium salt (2:1), Butyric acid, 2-hydroxy-4-(methylthio)-, calcium salt (2:1) (8CI), Calcium DL-2-hydroxy-4-methylthiobutyrate, Calcium DL-α-hydroxy-γ-methylthiobutyrate, Calcium α-hydroxy-γ-methylmercaptobutyra
InChI formel InChI=1S/2C5H10O3S.Ca/c2*1-9-3-2-4(6)5(7)8;/h2*4,6H,2-3H2,1H3,(H,7,8);/q;;+2/p-2
LEDER [Ca+2].CSCCC(O)C(=O)[O-].CSCCC(O)C(=O)[O-]
IUPAC-namn calcium;2-hydroxy-4-methylsulfanylbutanoate
Molekylvikt (g/mol) 338.454
Formel vikt 338.017