Läs mer
Calcium Ascorbate Dihydrate, TRC
Produktkod. 30347481
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30347481 1 g
1 options
Produktkod. 30347481 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC1450251G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ascorbic acid calcium salt dihydrate
CAS 5743-28-2
Molekylformel 2 C6 H7 O6 . Ca . 4 H2 O
Kvantitet 1 g
Synonym Calcium ascorbate, Calcium di[(R)-2-[(S)-1,2-dihydroxyethyl]-4-hydroxy-5-oxo-2H-furan-3-olate] dihydrate
InChI formel InChI=1S/2C6H8O6.Ca.4H2O/c2*7-1-2(8)5-3(9)4(10)6(11)12-5;;;;;/h2*2,5,7-10H,1H2;;4*1H2/q;;+2;;;;/p-2/t2*2-,5+;;;;;/m00...../s1
LEDER O.O.O.O.[Ca+2].OC[C@H](O)[C@H]1OC(=O)C(=C1[O-])O.OC[C@H](O)[C@H]2OC(=O)C(=C2[O-])O
IUPAC-namn calcium;(2R)-2-[(1S)-1,2-dihydroxyethyl]-4-hydroxy-5-oxo-2H-furan-3-olate;tetrahydrate
Molekylvikt (g/mol) 462.37
Formel vikt 462.0534