Läs mer
Calcium Butyrate (~90%), TRC
Produktkod. 30335598
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30335598 5 g
1 options
Produktkod. 30335598 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC1450455G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Calcium Butyrate
CAS 5743-36-2
Molekylformel C4H8Ca0.5O2
Kvantitet 5 g
Synonym Butanoic Acid, Calcium Salt, Butyric Acid, Calcium Salt, Greencab 70 Coated
InChI formel InChI=1S/2C4H8O2.Ca/c2*1-2-3-4(5)6;/h2*2-3H2,1H3,(H,5,6);/q;;+2/p-2
LEDER CCCC(=O)[O-].CCCC(=O)[O-].[Ca+2]
IUPAC-namn calcium;butanoate
Molekylvikt (g/mol) 108.14
Formel vikt 214.0518