Läs mer
Calcium Glycerophosphate Hydrate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Calcium Glycerophosphate Hydrate |
| CAS | 27214-00-2 |
| Molekylformel | C3H7CaO6P |
| Kvantitet | 5 g |
| Synonym | Calcium glycerophosphate, 1,2,3-Propanetriol, mono(dihydrogen phosphate), calcium salt (1:1), Calcium glycerophosphate (6CI,7CI), Glycerol, mono(dihydrogen phosphate), calcium salt (1:1) (8CI), Calcium glycerol phosphate, Nerozin, Neurosin, Prelief |
| InChI formel | InChI=1S/C3H9O6P.Ca/c4-1-3(5)2-9-10(6,7)8;/h3-5H,1-2H2,(H2,6,7,8);/q;+2/p-2 |
| LEDER | [Ca+2].[Ca+2].OCC(O)COP(=O)([O-])[O-].OCC(CO)OP(=O)([O-])[O-] |
| IUPAC-namn | calcium;2,3-dihydroxypropyl phosphate |
| Molekylvikt (g/mol) | 210.14 |
| Formel vikt | 419.9212 |