Learn More
Calcium ISE Standard Solution, Mettler Toledo™
Product Code. 11927521
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Product Code. Quantity Unit Size
11927521 500 mL 500mL
1 options
Product Code. 11927521 Supplier Mettler Toledo Online Supplier No. 51344771
  • ONLINE EXCLUSIVE PRICE
    1545.00 SEK
    1321.40 SEK / 500ml
    Save 223.60 SEK
    14% Av
 Restricted Item
I lager
Specifications
Description ISE standard Ca 1000 mg/L, 500mL
Packaging 500 mL
Physical Form Liquid
Color Colorless
Quantity 500 mL