Läs mer
Calcium-L-lactate Hydrate, TRC
Produktkod. 30331640
Change view
Kvantitet:
25 g
Produktkod. Kvantitet
30331640 25 g
1 options
Produktkod. 30331640 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC14568825G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Calcium-L-lactate Hydrate
CAS 949014-28-2
Molekylformel C6H10CaO6 . x(H2O)
Kvantitet 25 g
Synonym 2-Hydroxypropanoic Acid Calcium Salt Hydrate
InChI formel InChI=1S/C11H16BrO3P/c1-3-14-16(13,15-4-2)9-10-6-5-7-11(12)8-10/h5-8H,3-4,9H2,1-2H3
LEDER CCOP(=O)(Cc1cccc(Br)c1)OCC
IUPAC-namn 1-bromo-3-(diethoxyphosphorylmethyl)benzene
Molekylvikt (g/mol) 236.23
Formel vikt 306.002