Läs mer
Calcium Levofolinate, TRC
Produktkod. 30136445
Change view
Kvantitet:
5 mg
100 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136445 1 g
30008681 100 mg
30009831 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136445 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC1456351G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 80433-71-2
Molekylformel C20H23CaN7O7
Molekylvikt (g/mol) 513.13
Synonym N-[4-[[(2-Amino-5-formyl-1,4,5,6,7,8-hexahydro-4-oxo-6-pteridinyl)methyl]amino]benzoyl]-L-glutamic Acid Calcium Salt, N-[4-[[[(6S)-2-Amino-5-formyl-1,4,5,6,7,8-hexahydro-4-oxo-6-pteridinyl]methyl]amino]benzoyl]-L-glutamic Acid Calcium Salt, S)-Leucovorin Calcium Salt, 5-Formyl-(6S)-tetrahydrofolate calcium salt, CL 307782, Calcium (6S)-folinate Calcium levofolinate, Elvorine, Isovorin, L-Folinic Acid Calcium Salt, Levoleucovorin Calcium, (6S)-Folinic Acid Calcium Salt
IUPAC-namn calcium;(2S)-2-[[4-[(2-amino-5-formyl-4-oxo-3,6,7,8-tetrahydropteridin-6-yl)methylamino]benzoyl]amino]pentanedioate
LEDER [Ca+2].NC1=NC(=O)C2=C(NC[C@H](CNc3ccc(cc3)C(=O)N[C@@H](CCC(=O)[O-])C(=O)[O-])N2C=O)N1
Specifikationer
Kemiskt namn eller material Calcium Levofolinate
Kvantitet 1 g
InChI formel InChI=1S/C20H23N7O7.Ca/c21-20-25-16-15(18(32)26-20)27(9-28)12(8-23-16)7-22-11-3-1-10(2-4-11)17(31)24-13(19(33)34)5-6-14(29)30;/h1-4,9,12-13,22H,5-8H2,(H,24,31)(H,29,30)(H,33,34)(H4,21,23,25,26,32);/q;+2/p-2/t12?,13-;/m0./s1
Formel vikt 511.1128