Läs mer
Calcium polystyrene sulfonate, MedChemExpress
Produktkod. 16401099
Encompass_Preferred
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
16401099 1 g
1 options
Produktkod. 16401099 Leverantör Medchem Express Leverantörsnummer HY107929
MedChemExpress Calcium polystyrene sulfonate is an ion-exchange resin used for reducing blood levels of potassium. Calcium polystyrene sulfonate is used to treat hyperkalemia in patients with chronic kidney disease (CKD).
Calcium polystyrene sulfonate is an ion-exchange resin used for reducing blood levels of potassium. Calcium polystyrene sulfonate is used to treat hyperkalemia in patients with chronic kidney disease (CKD).
Specifikationer
Kemiskt namn eller material Calcium polystyrene sulfonate
Fysisk form Solid
CAS 37286-92-3
Färg White
Kvantitet 1 g
Anteckningar om renhetsgrad Research
Molekylformel (C8H8O3S)x.xCa
För användning med (applikation) Neuroscience-Neuromodulation
Synonym Poly(styrenesulfonic acid) calcium salt
Löslighetsinformation DMSO : 1 mg/mL (Need ultrasonic) ∣1M HCl : < 1 mg/mL (ultrasonic;adjust pH to 1 with HCl) (insoluble)
LEDER [Calcium polystyrene sulfonate]