missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CALD1 Mouse anti-Human, Mouse, Rat, Clone: 1E1C11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66693-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
Caldesmon is a developmentally regulated protein involved in smooth muscle and non-muscle contraction. Two closely related variants of human caldesmon have been identified which differ in their eletrophoretic mobility and cellular distribution. The h-caldesmon variant (120-150kDa) is predomitly expressed in smooth muscle whereas l-caldesmon (70-80kDa) is found in non-muscle tissue and cells. Neither of the two variants has been detected in skeletal muscle.Specifications
CALD1 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q05682, Q62736 | |
CALD1 | |
CALD1 Fusion Protein Ag13676 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG2b |
Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence | |
1E1C11 | |
Unconjugated | |
CALD1 | |
CAD, CALD1, Caldesmon, caldesmon 1, CDM, H CAD, L CAD, NAG22 | |
Mouse | |
Protein A | |
RUO | |
109624, 25687, 800 | |
-20°C | |
Liquid |