Läs mer
Calf thymus DNA, MedChemExpress
Produktkod. 16400136
Encompass_Preferred
Change view
Kvantitet:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16400136 2 mg
16494298 10 mg
16490627 50 mg
16487017 5 mg
16483026 25 mg
16415226 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Produktkod. 16400136 Leverantör Medchem Express Leverantörsnummer HY109517
MedChemExpress Calf thymus DNA (DNA from calf thymus) is high quality double-stranded template DNA isolated from the thymus of male and female calves.
Calf thymus DNA (DNA from calf thymus) is high quality double-stranded template DNA isolated from the thymus of male and female calves.
Specifikationer
Kemiskt namn eller material Calf thymus DNA
Fysisk form Powder
CAS 91080-16-9
Färg White
Kvantitet 2 mg
Anteckningar om renhetsgrad Research
För användning med (applikation) Cancer-programmed cell death
Synonym DNA from calf thymus, Thymonucleic acid
Löslighetsinformation H2O : 20 mg/mL (Need ultrasonic)
LEDER [Calf thymus DNA]
Procent renhet 95.0%