Läs mer
Calibration Mix 1 for AA and ICP-OES, Honeywell Fluka™
Produktkod. 16694194 Handla allt Honeywell Fluka Produkter
Change view
Kvantitet:
100 ml
Produktkod. Kvantitet
16694194 100 ml
1 options
Produktkod. 16694194 Leverantör Honeywell Fluka Leverantörsnummer E2020100ML
Request Bulk or Custom Format
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Honeywell Fluka™ analytical standards offer a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. The products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Calibration Mix 1 for AA and ICP-OES
Densitet 1.06 g/cm3
pH <2
Kvantitet 100 ml
Kvalitet Standards for ICP and ICP-MS