Learn More
WTW™ Calibration Vessel for WTW™ ConOx™ Conductivity/DO Combination Sensor
The calibration and storage containers for the WTW oxygen sensors are suitable for the standard conform calibration (DIN ISO 5814 or DIN ISO 17289: 2014-12) with water steam saturated air as well as for storage of the sensors to protect the sensitive membrane.
Brand: WTW™ 205400
Additional Details : Weight : 0.10000kg
Specifications
Calibration vessel | |
ConOx conductivity/DO combination sensor |