missing translation for 'onlineSavingsMsg'
Learn More

CALML5 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 13059-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16881084

  • 1236.20 SEK / 20µL
Estimated Shipment: 07-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a novel calcium binding protein expressed in the epidermis and related to the calmodulin family of calcium binding proteins. Functional studies with recombit protein demonstrate it does bind calcium and undergoes a conformational change when it does so. Abundant expression is detected only in reconstructed epidermis and is restricted to differentiating keratinocytes. In addition, it can associate with transglutaminase 3, shown to be a key enzyme in the terminal differentiation of keratinocytes.
Specifications

Specifications

CALML5
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9NZT1
CALML5, calmodulin like 5, Calmodulin like protein 5, Calmodulin like skin protein, CLSP
Rabbit
IgG
20 μL
-20°C
Primary
51806
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
CALML5
Liquid
CALML5
CALML5 Fusion Protein Ag3734
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
SDS
Documents
Certificates
Special Offers

Special Offers