missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ Calpain 1 Monoclonal Antibody (P-6(2H7))

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA113079
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
MA1-13079 detects calpain-1 large subunit (mu-type) in human and mouse samples by western blot, immunofluorescence, immunohistochemistry.
The calpains including Calpain 1 (CAPN1), calcium-activated neutral proteases, are nonlysosomal, intracellular cysteine proteases. The mammalian calpains include ubiquitous, stomach-specific, and muscle-specific proteins. The ubiquitous enzymes consist of heterodimers with distinct large, catalytic subunits associated with a common small, regulatory subunit. This gene encodes the large subunit of the ubiquitous enzyme, calpain 1. Several transcript variants encoding two different isoforms have been found for this gene. Diseases associated with CAPN1 include Spastic Paraplegia 76, Autosomal Recessive and Spasticity.
Specifikationer
| Calpain 1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide; pH 7.2-7.4 | |
| O35350, P07384 | |
| CAPN1 | |
| human erythrocyte Calpain-1 large subunit. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| P-6(2H7) | |
| Unconjugated | |
| CAPN1 | |
| calcium activated neutral protease; calcium-activated neutral protease 1; calcium-activated neutral proteinase 1; calcium-dependent protease; calpain; calpain 1; calpain 1 large subunit; calpain 1, (mu/I) large subunit; calpain 1, large subunit; calpain mu-type; calpain, large polypeptide L1; calpain-1 catalytic subunit; Calpain-1 large subunit; Calpain-I; Calpain-II; CANP; CANP 1; Canp1; CANPL1; Capa1; Capa-1; Capn1; Cell proliferation-inducing gene 30 protein; cell proliferation-inducing protein 30; Cls1; I79_024883; micromolar calcium-activated neutral protease 1; micromolar calcium-activated neutral protease 1 isoform A; micromolar calcium-activated neutral protease 1 isoform B; micromolar calcium-dependent neutral protease large subunit; micromolar-calpain; mucalpain; mu-calpain; mu-calpin; muCANP; muCL; PIG30; SPG76 | |
| Mouse | |
| Protein A | |
| RUO | |
| 12333, 823 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction