Learn More
Calpain 2 Monoclonal Antibody (107-82), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA3942
3228.00 SEK valid until 2024-06-28
Use promo code "21649" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
MA3-942 detects the 80 kDa subunit of m-calpain in human platelets and erythrocytes, bovine platelets, heart and skeletal muscle, rat myoblasts, kidney, liver and spleen, pig cultured cells, and rabbit samples. This antibody does not cross-react with mu-calpain, n-calpain, calmodulin or calpastatin. MA3-942 has been successfully used in Western blot, IF, immunohistochemistry, and ELISA procedures. By Western blot, this antibody detects an ∽80 kDa protein representing m-calpain in bovine platelets. Immunocytochemical staining of m-calpain in LLC-PK1 cells with MA3-942 results in diffuse cytoplasmic staining. This product has not been shown to be effective in immunoprecipitation experiments. The MA3-942 antigen is purified bovine skeletal muscle 80 kDa subunit of m-calpain. This antibody recognizes an epitope between amino acids 502-699 (domain III/IV) of human calpain.
Mu-Calpain, also known as Calpain-I, and m-calpain, also known as Calpain-II, are intracellular, calcium-dependent cysteine proteases. Mu-calpain has a micromolar sensitivity (thus the mu-) as compared to the millimolar calcium sensitivity of m-calpain.Specifications
Calpain 2 | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
P06814, P17655, P43367, Q07009, Q27971 | |
CAPN2 | |
Purified bovine skeletal muscle 80 kDa subunit of m-calpain. | |
100 μg | |
Primary | |
Bovine, Human, Porcine, Rabbit, Rat | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot | |
107-82 | |
Unconjugated | |
CAPN2 | |
80 kDa M-calpain subunit; AI326419; Calcium-activated neutral proteinase 2; calcium-dependent protease; CALP80; calpain 2; calpain 2, (m/II) large subunit; calpain 2, large [catalytic] subunit; calpain 2, large subunit; calpain II; calpain large polypeptide L2; Calpain M-type; calpain, large polypeptide L2; calpain-2 catalytic subunit; Calpain-2 large subunit; CANP 2; CANP2; CANPL2; CANPml; Capa2; Capa-2; CAPN2; cysteine proteinase; m-calpain; m-calpain 80 kDa large subunit; m-calpain large subunit; m-calpin; mCANP; millimolar-calpain | |
Mouse | |
Protein A | |
RUO | |
100009092, 281662, 29154, 397393, 824 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |