missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CAMK1 (CaMK1) Recombinant Human Protein

CAMK1 (CaMK1) is a serine/threonine kinase that is a member of the calcium/calmodulin-dependent protein kinase family.

Brand:  Invitrogen™ PR8013A

Additional Details : Weight : 0.01000kg

Product Code. 15408643

  • 18240.00 SEK / 100µg
Estimated Shipment: 30-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CAMK1 (CaMK1) is involved in cell differentiation, nervous system development, and signal transduction.

Specifications

Specifications

NP_003547
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol
100μg
RUO - research use only
CAMK1 (CaMK1)
Human
His Tag
Frozen
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology
45.4kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
CAMK1 Kinase Family
Recombinant
Baculovirus
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers