missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CAMK4 (Human) Recombinant Protein
Human CAMK4 full-length ORF ( AAH25687.1, 1 a.a. - 473 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00000814-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MLKVTVPSCSASSCSSVTASAAPGTASLVPDYWIDGSNRDALSDFFEVESELGRGATSIVYRCKQKGTQKPYALKVLKKTVDKKIVRTEIGVLLRLSHPNIIKLKEIFETPTEISLVLELVTGGELFDRIVEKGYYSERDAADAVKQILEAVAYLHENGIVHRDLKPENLLYATPAPDAPLKIADFGLSKIVEHQVLMKTVCGTPGYCAPEILRGCAYGPEVDMWSVGIITYILLCGFEPFYDERGDQFMFRRILNCEYYFISPWWDEVSLNAKDLVRKLIVLDPKKRLTTFQALQHPWVTGKAANFVHMDTAQKKLQEFNARRKLKAAVKAVVASSRLGSASSSHGSIQESHKASRDPSPIQDGNEDMKAIPEGEKIQGDGAQAAVKGAQAELMKVQALEKVKGADINAEEAPKMVPKAVEDGIKVADLELEEGLAEEKLKTVEEAAAPREGQGSSAVGFEVPQQDVILPEY
Specifications
AAH25687.1 | |
calcium/calmodulin-dependent protein kinase IV | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CAMK4 | |
GST |
814 | |
Wheat germ expression system | |
25 μg | |
CaMK-GR, MGC36771 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |