missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CaMKII alpha Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1236.20 SEK - 3313.85 SEK
Specifications
Antigen | CaMKII alpha |
---|---|
Concentration | 0.32 mg/mL |
Applications | Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16850245
|
Proteintech
20666-1-AP-20UL |
20 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16840245
|
Proteintech
20666-1-AP-150UL |
150 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
CaM kinase II, in a calcium and calmodulin dependent manner, phosphorylates many different brain substrates including synapsin I, tryptophan hydroxylase, tyrosine hydroxylase and nitric oxide synthase thereby performing regulatory functions associated with increases in intracellular free calcium. CaM kinase II is particularly abundant in the hippocampus and forebrain where it comprises approximately 1% of total protein. Within the neuron, CaM kinase II is a major component of the postsynaptic density fraction accounting for as much as 30% of total protein in the post synaptic density (PSD) region. Autophosphorylation of threonine-286 occurs after calcium/calmodulin activation and can enable CaM kinase II to remain active independent of free calcium. Autophosphorylation of CaM kinase II in hippocampal neurons, which results in relatively high levels of kinase activity even at basal calcium concentrations, may allow for both upward and downward regulation as opposed to simply on/off regulation.Specifications
CaMKII alpha | |
Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human | |
Q9UQM7 | |
815 | |
Peptide | |
Primary | |
-20°C | |
CAMK2A |
0.32 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CaM kinase II subunit alpha, CaMK II subunit alpha, CAMK2A, CAMKA, CaMKII alpha, KIAA0968 | |
CAMK2A | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |