missing translation for 'onlineSavingsMsg'
Learn More

CaMKII beta Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 55218-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16826715

  • 1236.20 SEK / 20µL
Estimated Shipment: 07-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This antibody can recognized all the isoform except 7 of CaMKII beta.

The Ca2+/calmodulin-dependent protein kinases (CaM kinases) comprise a structurally related subfamily of serine/threonine kinases which include CaMKI, CaMKII and CaMKIV. CaMKII is an ubiquitously expressed serine/threonine protein kinase that is activated by Ca2+ and calmodulin (CaM) and has been implicated in regulation of the cell cycle and transcription. There are four CaMKII isozymes, designated alpha, beta, gamma and delta, which may or may not be co-expressed in the same tissue type. CaMKII alpha is autophosphorylated on Thr 286 upon the binding of the Ca2+/CaM complex to the autoinhibitory domain of CaMKII. This process is called Ca2+/CaM trapping, which is thought to be involved in the synaptic encoding of information.
Specifications

Specifications

CaMKII beta
0.28 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P28652, Q13554
CaM kinase II subunit beta, CAM2, CaMK II subunit beta, CAMK2, CAMK2B, CAMKB, CaMKII beta
Rabbit
IgG
20 μL
-20°C
Primary
12323, 24245, 816
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
CAMK2B
Liquid
CAMK2B
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Rat, Mouse
Videos
SDS
Documents
Certificates
Special Offers

Special Offers