Läs mer
Campesterol, TRC
Produktkod. 17910055
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17910055 10 mg
17909424 5 mg
17948194 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17910055 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC15536010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 474-62-4
Molekylformel C28 H48 O
Molekylvikt (g/mol) 400.68
Synonym (3beta,24R)-Ergost-5-en-3-ol, Campesterin, (24R)-Methylcholest-5-en-3beta-ol, 24(R)-Methylcholesterol, 24alpha-Methyl-5-cholesten-3beta-ol, 24alpha-Methylcholesterol, delta5-24-Isoergosten-3beta-ol, NSC 224330;
IUPAC-namn (3S,8S,9S,10R,13R,14S,17R)-17-[(2R,5S)-5,6-dimethylheptan-2-yl]-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
LEDER CC(C)[C@@H](C)CC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
Specifikationer
Kemiskt namn eller material Campesterol
Anteckningar om renhetsgrad GC
Kvantitet 10 mg
InChI formel InChI=1S/C28H48O/c1-18(2)19(3)7-8-20(4)24-11-12-25-23-10-9-21-17-22(29)13-15-27(21,5)26(23)14-16-28(24,25)6/h9,18-20,22-26,29H,7-8,10-17H2,1-6H3/t19-,20+,22-,23-,24+,25-,26-,27-,28+/m0/s1
Formel vikt 400.3705
Procent renhet >95%