Läs mer
Canadine, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Tetrahydroberberine (Canadine) |
| CAS | 522-97-4 |
| Molekylformel | C20 H21 N O4 |
| Kvantitet | 10 mg |
| Synonym | 6H-Benzo[g]-1,3-benzodioxolo[5,6-a]quinolizine, 5,8,13,13a-tetrahydro-9,10-dimethoxy-, Berberine, 9-deoxy-16,17-dihydro- (6CI), Berbine, 9,10-dimethoxy-2,3-(methylenedioxy)- (7CI,8CI), (±)-Canadine, (±)-Tetrahydroberberine, Berberine, tetrahydro-, Canadine, NSC 36351, NSC 94918, Tetrahydroberberine, Tetrahydroumbellatine, Xanthopuccine, dl-Canadine, dl-Tetrahydroberberine |
| InChI formel | InChI=1S/C20H21NO4/c1-22-17-4-3-12-7-16-14-9-19-18(24-11-25-19)8-13(14)5-6-21(16)10-15(12)20(17)23-2/h3-4,8-9,16H,5-7,10-11H2,1-2H3 |
| LEDER | COc1ccc2CC3N(CCc4cc5OCOc5cc34)Cc2c1OC |
| IUPAC-namn | 16,17-dimethoxy-5,7-dioxa-13-azapentacyclo[11.8.0.02,10.04,8.015,20]henicosa-2,4(8),9,15(20),16,18-hexaene |
| Molekylvikt (g/mol) | 339.385 |
| Formel vikt | 339.1471 |