Läs mer
Canagliflozin Hydrate, TRC
Produktkod. 30335055
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Kvantitet:
1 mg
Product Code. Kvantitet
30335055 1 mg
1 missing translation for 'options'
Product Code. 30335055 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCC1752591MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material Canagliflozin Hydrate
CAS 1869942-80-2
Molekylformel C24 H25 F O5 S . x(H2O)
Kvantitet 1 mg
InChI formel InChI=1S/C24H25FO5S.H2O/c1-13-2-3-15(24-23(29)22(28)21(27)19(12-26)30-24)10-16(13)11-18-8-9-20(31-18)14-4-6-17(25)7-5-14;/h2-10,19,21-24,26-29H,11-12H2,1H3;1H2/t19-,21-,22+,23-,24+;/m1./s1
LEDER O.Cc1ccc(cc1Cc2ccc(s2)c3ccc(F)cc3)[C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O
IUPAC-namn (2S,3R,4R,5S,6R)-2-[3-[[5-(4-fluorophenyl)thiophen-2-yl]methyl]-4-methylphenyl]-6-(hydroxymethyl)oxane-3,4,5-triol;hydrate
Molekylvikt (g/mol) 462.5