Läs mer
Candesartan Cilexetil, TRC
Produktkod. 30007960
Change view
Kvantitet:
50 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30007960 1 g
18021950 50 mg
18041050 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30007960 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC1755801G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 145040-37-5
Molekylformel C33 H34 N6 O6
Molekylvikt (g/mol) 610.66
Synonym Candesartan Cilexetil, 2-Ethoxy-1-[[2'-(2H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-1H-benzimidazole-7-carboxylic acid 1-[[(cyclohexyloxy)carbonyl]oxy]ethyl ester, (±)-2-Ethoxy-1-[[2'-(2H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-1H-benzimidazole-7-carboxylic acid 1-[[(cyclohexyloxy)carbonyl]oxy]ethyl ester, (±)-1-Hydroxyethyl 2-ethoxy-1-[p-(o-1H-tetrazol-5-ylphenyl)l]-7-benzimidazolecarboxylate, cyclohexyl carbonate (ester), 1-[[(Cyclohexyloxy)carbonyl]oxy]ethyl 2-ethoxy-1-[[2'-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl]benzimidazole-7-carboxylate, 2-Ethoxy-1-[(2'-(1H-tetrazol-5-yl)-1,1'-biphenyl-4-yl)methyl]-7-benzimidazolecarboxylic acid 1-(cyclohexyloxycarbonyloxy)ethyl ester, Atacand, Blopress, Candesartan cilexetil, TCV 116, TCY 116
IUPAC-namn 1-cyclohexyloxycarbonyloxyethyl 2-ethoxy-3-[[4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methyl]benzimidazole-4-carboxylate
LEDER CCOc1nc2cccc(C(=O)OC(C)OC(=O)OC3CCCCC3)c2n1Cc4ccc(cc4)c5ccccc5c6nnn[nH]6
Specifikationer
Kemiskt namn eller material Candesartan cilexetil
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C33H34N6O6/c1-3-42-32-34-28-15-9-14-27(31(40)43-21(2)44-33(41)45-24-10-5-4-6-11-24)29(28)39(32)20-22-16-18-23(19-17-22)25-12-7-8-13-26(25)30-35-37-38-36-30/h7-9,12-19,21,24H,3-6,10-11,20H2,1-2H3,(H,35,36,37,38)
Formel vikt 610.254
Procent renhet >95%