Läs mer
Candesartan Ethyl Ester, TRC
Produktkod. 17912615
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17912615 10 mg
17952805 5 mg
17914945 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17912615 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC17557010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 139481-58-6
Molekylformel C26 H24 N6 O3
Molekylvikt (g/mol) 468.51
Synonym 1H-Benzimidazole-7-carboxylic acid, 2-ethoxy-1-[[2'-(2H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-, ethyl ester, 1H-Benzimidazole-7-carboxylic acid, 2-ethoxy-1-[[2'-(1H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-, ethyl ester (9CI), Ethyl 2-ethoxy-1-[[2'-(2H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-1H-benzimidazole-7-carboxylate, Ethyl 2-Ethoxy-1-[[2'-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl]-1H-benzimidazole-7-carboxylate, Candesartan Cilexetil Imp. A (EP), Candesartan Ethyl Ester
IUPAC-namn ethyl 2-ethoxy-3-[[4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methyl]benzimidazole-4-carboxylate
LEDER CCOC(=O)c1cccc2nc(OCC)n(Cc3ccc(cc3)c4ccccc4c5nnn[nH]5)c12
Specifikationer
Kemiskt namn eller material Candesartan ethyl ester
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C26H24N6O3/c1-3-34-25(33)21-10-7-11-22-23(21)32(26(27-22)35-4-2)16-17-12-14-18(15-13-17)19-8-5-6-9-20(19)24-28-30-31-29-24/h5-15H,3-4,16H2,1-2H3,(H,28,29,30,31)
Formel vikt 468.191
Procent renhet >95%