Läs mer
Candesartan Methyl Ester, TRC
Produktkod. 30137298
Change view
Kvantitet:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137298 25 mg
30003347 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30137298 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC17559525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 139481-69-9
Molekylformel C25 H22 N6 O3
Molekylvikt (g/mol) 454.4806
Synonym 1H-Benzimidazole-7-carboxylic acid, 2-ethoxy-1-[[2'-(2H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-, methyl ester, 1H-Benzimidazole-7-carboxylic acid, 2-ethoxy-1-[[2'-(1H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-, methyl ester (9CI), Methyl 1-[(2'-(1H-tetrazol-5-yl)biphenyl-4-yl)methyl]-2-ethoxybenzimidazole-7-carboxylate, Methyl 2-ethoxy-1-[[2'-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl]benzimidazole-7-carboxylate, Candesartan Cilexetil Imp. I (EP)
IUPAC-namn methyl 2-ethoxy-3-[[4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methyl]benzimidazole-4-carboxylate
LEDER CCOc1nc2cccc(C(=O)OC)c2n1Cc3ccc(cc3)c4ccccc4c5nnn[nH]5
Specifikationer
Kemiskt namn eller material Candesartan Methyl Ester
Kvantitet 25 mg
InChI formel InChI=1S/C25H22N6O3/c1-3-34-25-26-21-10-6-9-20(24(32)33-2)22(21)31(25)15-16-11-13-17(14-12-16)18-7-4-5-8-19(18)23-27-29-30-28-23/h4-14H,3,15H2,1-2H3,(H,27,28,29,30)
Formel vikt 454.1753